4-Fluoro-2-hydroxybenzonitrile 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Cyano-5-fluorophenol, 4-Fluorosalicylonitrile

PC51150-5G
APOSPC51150-5GEA 150 EUR
APOSPC51150-5G
4-Fluoro-2-hydroxybenzonitrile 98%
4-Fluoro-2-hydroxybenzonitrile
Formel: C₇H₄FNO
Molecular Weight: 137,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185487
CAS-Nummer: 186590-01-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR