2,6-Difluoro-4-hydroxybenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Difluoro-4-(hydroxymethyl)phenol

PC51152-1G
APOSPC51152-1GEA 40.6 EUR
APOSPC51152-1G
2,6-Difluoro-4-hydroxybenzyl alcohol 95%
2,6-Difluoro-4-hydroxybenzyl alcohol
Formel: C₇H₆F₂O₂
Schmelzpunkt: 160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11040173
CAS-Nummer: 438049-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR