3,4-Dihydro-6-(trifluoromethyl)-2H-1,4-benzoxazine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dihydro-6-(trifluoromethyl)-2H-benzo[b][1,4]oxazine

PC51155-1G
APOSPC51155-1GEA 0 EUR
APOSPC51155-1G
3,4-Dihydro-6-(trifluoromethyl)-2H-1,4-benzoxazine 97%
3,4-Dihydro-6-(trifluoromethyl)-2H-1,4-benzoxazine
Formel: C₉H₈F₃NO
Molecular Weight: 203,161 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08544345
CAS-Nummer: 347-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR