2'-Fluoro-3'-(trifluoromethoxy)acetophenone 97%

Lieferant: Apollo Scientific
Achtung

PC51175-1G PC51175-250MG PC51175-5G
APOSPC51175-1GEA 0 EUR
APOSPC51175-1G APOSPC51175-250MG APOSPC51175-5G
2'-Fluoro-3'-(trifluoromethoxy)acetophenone 97%
2'-Fluoro-3'-(trifluoromethoxy)acetophenone
Formel: C₉H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23703636
CAS-Nummer: 1429321-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR