2,4-Difluoro-3-(trifluoromethyl)benzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: α,α,α,2,4-Pentafluoro-m-tolualdehyde, 2,6-Difluoro-3-formylbenzotrifluoride

PC51182-1G PC51182-250MG PC51182-5G
APOSPC51182-1GEA 101 EUR
APOSPC51182-1G APOSPC51182-250MG APOSPC51182-5G
2,4-Difluoro-3-(trifluoromethyl)benzaldehyde 98%
2,4-Difluoro-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₃F₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD12404958
CAS-Nummer: 134099-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR