3'-(Pentafluorthio)acetophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC51188-1G
PC51188-5G
APOSPC51188-1GEA
429
EUR
APOSPC51188-1G
APOSPC51188-5G
3'-(Pentafluorthio)acetophenon 97%
3'-(Pentafluorthio)acetophenon
Formel:
C₈H₇F₅OS Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652407 CAS-Nummer: 159896-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...