5-(Trifluoromethyl)benzene-1,3-diboronic acid 98%

Lieferant: Apollo Scientific
Achtung

PC5119-1G
APOSPC5119-1GEA 176 EUR
APOSPC5119-1G
5-(Trifluoromethyl)benzene-1,3-diboronic acid 98%
5-(Trifluoromethyl)benzene-1,3-diboronic acid
Formel: C₇H₇B₂F₃O₄
Molecular Weight: 233,745 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08235085
CAS-Nummer: 913835-35-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR