4-Bromo-6-(pentafluorothio)-1H-benzimidazole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Bromo-1H-benzimidazol-6-yl)sulphur pentafluoride, 4-Bromo-6-(pentafluorosulphanyl)-1H-benzimidazole

PC51190-1G PC51190-250MG
APOSPC51190-1GEA 429 EUR
APOSPC51190-1G APOSPC51190-250MG
4-Bromo-6-(pentafluorothio)-1H-benzimidazole 95%
4-Bromo-6-(pentafluorothio)-1H-benzimidazole
Formel: C₇H₄BrF₅N₂S
Molecular Weight: 323.081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369706
CAS-Nummer: 1432075-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR