2-(Difluoromethyl)-4-hydroxypyridine 95%

Lieferant: Apollo Scientific
Achtung

PC51200-1G PC51200-250MG
APOSPC51200-1GEA 0 EUR
APOSPC51200-1G APOSPC51200-250MG
2-(Difluoromethyl)-4-hydroxypyridine 95%
2-(Difluoromethyl)-4-hydroxypyridine
Formel: C₆H₅F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18820876
CAS-Nummer: 501125-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR