2-(Difluoromethyl)isonicotinonitrile

Lieferant: Apollo Scientific
Achtung

PC51202-1G PC51202-250MG
APOSPC51202-1GEA 0 EUR
APOSPC51202-1G APOSPC51202-250MG
2-(Difluoromethyl)isonicotinonitrile
2-(Difluoromethyl)isonicotinonitrile
Formel: C₇H₄F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18256051
CAS-Nummer: 1211528-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR