2-(Difluoromethyl)isonicotinaldehyde

Lieferant: Apollo Scientific
Achtung

PC51203-100MG
APOSPC51203-100MGEA 205 EUR
APOSPC51203-100MG
2-(Difluoromethyl)isonicotinaldehyde
2-(Difluoromethyl)isonicotinaldehyde
Formel: C₇H₅F₂NO
Lagertemperatur: Kühlschrank
MDL: MFCD18256101
CAS-Nummer: 1211540-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR