2-(Difluoromethyl)isonicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Difluoromethyl)pyridine-4-carboxylic acid

PC51206-250MG PC51206-50MG
APOSPC51206-250MGEA 234 EUR
APOSPC51206-250MG APOSPC51206-50MG
2-(Difluoromethyl)isonicotinic acid
2-(Difluoromethyl)isonicotinic acid
Formel: C₇H₅F₂NO₂
Molecular Weight: 173.117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18256212
CAS-Nummer: 1256818-14-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR