Methyl 4-chloro-6-(trifluoromethyl)nicotinate

Lieferant: Apollo Scientific
Achtung

PC51211-1G PC51211-250MG
APOSPC51211-1GEA 0 EUR
APOSPC51211-1G APOSPC51211-250MG
Methyl 4-chloro-6-(trifluoromethyl)nicotinate
Methyl 4-chloro-6-(trifluoromethyl)nicotinate
Formel: C₈H₅ClF₃NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD18258301
CAS-Nummer: 1211539-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR