4-Chloro-5-nitro-2-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-5-nitro-α,α,α-trifluoro-2-picoline

PC51213-250MG
APOSPC51213-250MGEA 488 EUR
APOSPC51213-250MG
4-Chloro-5-nitro-2-(trifluoromethyl)pyridine
4-Chloro-5-nitro-2-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₃N₂O₂
Molecular Weight: 226,54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848352
CAS-Nummer: 438554-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR