2,4-Difluoro-5-nitrobenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC51227-1G PC51227-250MG PC51227-5G
APOSPC51227-1GEA 328 EUR
APOSPC51227-1G APOSPC51227-250MG APOSPC51227-5G
2,4-Difluoro-5-nitrobenzaldehyde 95%
2,4-Difluoro-5-nitrobenzaldehyde
Formel: C₇H₃F₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11857779
CAS-Nummer: 127228-77-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR