(Trifluoromethyl)cyclohexane

Lieferant: Apollo Scientific
Achtung

PC51228-1G PC51228-5G
APOSPC51228-1GEA 83.2 EUR
APOSPC51228-1G APOSPC51228-5G
(Trifluoromethyl)cyclohexane
(Trifluoromethyl)cyclohexane
Formel: C₇H₁₁F₃
Siedepunkt: 104…107 °C
Dichte: 1,149 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD03412289
CAS-Nummer: 401-75-2
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR