4-(Difluoromethoxy)-3-fluorobenzyl bromide ≥95%

Lieferant: Apollo Scientific

PC51313-250MG PC51313-1G PC51313-5G
APOSPC51313-250MGEA 208 EUR
APOSPC51313-250MG APOSPC51313-1G APOSPC51313-5G
4-(Difluoromethoxy)-3-fluorobenzyl bromide ≥95%
4-(Difluoromethoxy)-3-fluorobenzyl bromide
Formel: C₈H₆BrF₃O
MDL: MFCD18394990
CAS-Nummer: 181806-49-5
UN: 3265

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR