Methyl pentafluorobut-3-enoate 94%

Lieferant: Apollo Scientific
Gefahr

PC5134L-1G
APOSPC5134L-1GEA 415 EUR
APOSPC5134L-1G
Methyl pentafluorobut-3-enoate 94%
Methyl pentafluorobut-3-enoate
Formel: C₅H₃F₅O₂
Molecular Weight: 190,068 g/mol
Siedepunkt: 94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00054725
CAS-Nummer: 20562-79-2
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR