3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC51362-1G
PC51362-250MG
APOSPC51362-1GEA
0
EUR
APOSPC51362-1G
APOSPC51362-250MG
3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid
3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid
Formel:
C₁₀H₈ClF₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393906 CAS-Nummer: 1261873-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...