3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid

Lieferant: Apollo Scientific
Achtung

PC51362-1G PC51362-250MG
APOSPC51362-1GEA 0 EUR
APOSPC51362-1G APOSPC51362-250MG
3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid
3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid
Formel: C₁₀H₈ClF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18393906
CAS-Nummer: 1261873-29-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR