2-Chloro-3-iodo-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC51380-1G PC51380-250MG PC51380-5G
APOSPC51380-1GEA 429 EUR
APOSPC51380-1G APOSPC51380-250MG APOSPC51380-5G
2-Chloro-3-iodo-6-(trifluoromethyl)pyridine
2-Chloro-3-iodo-6-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₃IN
Schmelzpunkt: 35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD15478058
CAS-Nummer: 205240-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR