1,5-Dibromo-2,3,4-trifluorobenzene

Lieferant: Apollo Scientific

PC51386-250MG
APOSPC51386-250MGEA 0 EUR
APOSPC51386-250MG
1,5-Dibromo-2,3,4-trifluorobenzene
1,5-Dibromo-2,3,4-trifluorobenzene
Formel: C₆HBr₂F₃
Molecular Weight: 289.875 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28505052
CAS-Nummer: 17299-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR