tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate 97%

Lieferant: Apollo Scientific

PC51388-1G
APOSPC51388-1GEA 357 EUR
APOSPC51388-1G
tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate 97%
tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate
Formel: C₉H₁₆F₂N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD23105898
CAS-Nummer: 1408074-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR