3,4-Diaminophenylsulphur pentafluoride 97%

Lieferant: Apollo Scientific
Achtung

PC5149-1G
APOSPC5149-1GEA 348 EUR
APOSPC5149-1G
3,4-Diaminophenylsulphur pentafluoride 97%
3,4-Diaminophenylsulphur pentafluoride
Formel: C₆H₇F₅N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11501040
CAS-Nummer: 663179-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR