3-Hydroxy-5-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC51525-1G PC51525-250MG
APOSPC51525-1GEA 415 EUR
APOSPC51525-1G APOSPC51525-250MG
3-Hydroxy-5-(trifluoromethyl)benzyl alcohol
3-Hydroxy-5-(trifluoromethyl)benzyl alcohol
Formel: C₈H₇F₃O₂
Molecular Weight: 192.135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18397635
CAS-Nummer: 1261611-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR