5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine 97%

Lieferant: Apollo Scientific
Achtung

PC5169-1G PC5169-250MG PC5169-5G
APOSPC5169-1GEA 198 EUR
APOSPC5169-1G APOSPC5169-250MG APOSPC5169-5G
5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine 97%
5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine
Formel: C₇H₆F₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03094268
CAS-Nummer: 179062-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR