N-Methyl-2-(trifluormethoxy)anilin

Lieferant: Apollo Scientific
Achtung

PC5171E-1G
APOSPC5171E-1GEA 119 EUR
APOSPC5171E-1G
N-Methyl-2-(trifluormethoxy)anilin
N-Methyl-2-(trifluormethoxy)anilin
Formel: C₈H₈F₃NO
Molecular Weight: 191.15 g/mol
Siedepunkt: 74…76 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00052334
CAS-Nummer: 175278-04-3
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR