2,5-Difluorobenzene-1,4-diamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Difluorophenylene-1,4-diamine, 1,4-Diamino-2,5-difluorobenzene

PC5189-500MG
APOSPC5189-500MGEA 205 EUR
APOSPC5189-500MG
2,5-Difluorobenzene-1,4-diamine
2,5-Difluorobenzene-1,4-diamine
Formel: C₆H₆F₂N₂
Molecular Weight: 144,122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094275
CAS-Nummer: 698-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR