Ethyl 5-amino-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520000-1G
APOSPC520000-1GEA 797 EUR
APOSPC520000-1G
Ethyl 5-amino-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate
Formel: C₈H₁₀F₃N₃O₂
Molecular Weight: 237,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135205
CAS-Nummer: 866837-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR