Ethyl 5-amino-4-chloro-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520001-1G
APOSPC520001-1GEA 912 EUR
APOSPC520001-1G
Ethyl 5-amino-4-chloro-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate
Formel: C₈H₉ClF₃N₃O₂
Molecular Weight: 271,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135206
CAS-Nummer: 1427011-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR