Ethyl 5-amino-4-iodo-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520003-1G
APOSPC520003-1GEA 1140 EUR
APOSPC520003-1G
Ethyl 5-amino-4-iodo-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate
Formel: C₈H₉F₃IN₃O₂
Molecular Weight: 363,076 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135208
CAS-Nummer: 1427013-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR