Ethyl 5-amino-4-chloro-1-(4-fluorophenyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
PC520004-1G
APOSPC520004-1GEA
912
EUR
APOSPC520004-1G
Ethyl 5-amino-4-chloro-1-(4-fluorophenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(4-fluorophenyl)pyrazole-3-carboxylate
Formel:
C₁₂H₁₁ClFN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135136 CAS-Nummer: 1427011-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...