Ethyl 5-amino-1-(4-fluorophenyl)-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520006-1G
APOSPC520006-1GEA 1030 EUR
APOSPC520006-1G
Ethyl 5-amino-1-(4-fluorophenyl)-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-(4-fluorophenyl)-4-iodo-pyrazole-3-carboxylate
Formel: C₁₂H₁₁FIN₃O₂
Molecular Weight: 375,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135138
CAS-Nummer: 1427011-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR