Ethyl 5-amino-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520007-1G
APOSPC520007-1GEA 797 EUR
APOSPC520007-1G
Ethyl 5-amino-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Formel: C₁₃H₁₂F₃N₃O₂
Molecular Weight: 299,248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07388458
CAS-Nummer: 866838-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR