Ethyl 5-amino-4-chloro-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Lieferant: Apollo Scientific
PC520008-1G
APOSPC520008-1GEA
912
EUR
APOSPC520008-1G
Ethyl 5-amino-4-chloro-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Formel:
C₁₃H₁₁ClF₃N₃O₂ Molecular Weight: 333,694 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135139 CAS-Nummer: 1427023-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...