Ethyl 5-amino-1-[2-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520012-1G
APOSPC520012-1GEA 1320 EUR
APOSPC520012-1G
Ethyl 5-amino-1-[2-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-1-[2-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Formel: C₁₃H₁₂F₃N₃O₃
Molecular Weight: 315,248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135142
CAS-Nummer: 1427010-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR