Ethyl 5-amino-1-[3-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Lieferant: Apollo Scientific
PC520013-1G
APOSPC520013-1GEA
1320
EUR
APOSPC520013-1G
Ethyl 5-amino-1-[3-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-1-[3-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Formel:
C₁₃H₁₂F₃N₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135143 CAS-Nummer: 1427013-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...