Ethyl 5-amino-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520014-1G
APOSPC520014-1GEA 1320 EUR
APOSPC520014-1G
Ethyl 5-amino-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Formel: C₁₃H₁₂F₃N₃O₃
Molecular Weight: 315.248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07388473
CAS-Nummer: 1369271-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR