Ethyl 5-amino-4-chloro-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Lieferant: Apollo Scientific
PC520015-1G
APOSPC520015-1GEA
1030
EUR
APOSPC520015-1G
Ethyl 5-amino-4-chloro-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Formel:
C₁₃H₁₁ClF₃N₃O₃ Molecular Weight: 349.693 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135144 CAS-Nummer: 1427011-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...