Ethyl 5-amino-4-bromo-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520016-1G
APOSPC520016-1GEA 1030 EUR
APOSPC520016-1G
Ethyl 5-amino-4-bromo-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Formel: C₁₃H₁₁BrF₃N₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD23135145
CAS-Nummer: 1427021-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR