Ethyl 5-amino-4-iodo-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Lieferant: Apollo Scientific
PC520017-1G
APOSPC520017-1GEA
1140
EUR
APOSPC520017-1G
Ethyl 5-amino-4-iodo-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Formel:
C₁₃H₁₁F₃IN₃O₃ Molecular Weight: 441.144 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135146 CAS-Nummer: 1427023-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...