7-Bromo-2-(difluoromethyl)quinoline

Lieferant: Apollo Scientific

PC520018-1G
APOSPC520018-1GEA 3190 EUR
APOSPC520018-1G
7-Bromo-2-(difluoromethyl)quinoline
7-Bromo-2-(difluoromethyl)quinoline
Formel: C₁₀H₆BrF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD29767503
CAS-Nummer: 2091775-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR