4-[6-(Difluoromethyl)-2-pyridyl]aniline

Lieferant: Apollo Scientific

PC520020-1G
APOSPC520020-1GEA 2740 EUR
APOSPC520020-1G
4-[6-(Difluoromethyl)-2-pyridyl]aniline
4-[6-(Difluoromethyl)-2-pyridyl]aniline
Formel: C₁₂H₁₀F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29767505
CAS-Nummer: 2092578-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR