8-Bromo-2-(difluoromethyl)quinoline
Lieferant: Apollo Scientific
PC520023-1G
APOSPC520023-1GEA
3190
EUR
APOSPC520023-1G
8-Bromo-2-(difluoromethyl)quinoline
8-Bromo-2-(difluoromethyl)quinoline
Formel:
C₁₀H₆BrF₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767507 CAS-Nummer: 2091004-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...