tert-Butyl 2-[6-chloro-2-(difluoromethyl)-3-pyridyl]acetate

Lieferant: Apollo Scientific

PC520024-1G
APOSPC520024-1GEA 3190 EUR
APOSPC520024-1G
tert-Butyl 2-[6-chloro-2-(difluoromethyl)-3-pyridyl]acetate
tert-Butyl 2-[6-chloro-2-(difluoromethyl)-3-pyridyl]acetate
Formel: C₁₂H₁₄ClF₂NO₂
Molecular Weight: 277,695 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767508
CAS-Nummer: 2149590-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR