3-Bromo-6-(difluoromethyl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

PC520027-1G
APOSPC520027-1GEA 3190 EUR
APOSPC520027-1G
3-Bromo-6-(difluoromethyl)imidazo[1,2-a]pyridine
3-Bromo-6-(difluoromethyl)imidazo[1,2-a]pyridine
Formel: C₈H₅BrF₂N₂
Molecular Weight: 247,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767511
CAS-Nummer: 2090187-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR