tert-Butyl N-[3-(difluoromethyl)-4-pyridyl]carbamate

Lieferant: Apollo Scientific

PC520029-1G
APOSPC520029-1GEA 3190 EUR
APOSPC520029-1G
tert-Butyl N-[3-(difluoromethyl)-4-pyridyl]carbamate
tert-Butyl N-[3-(difluoromethyl)-4-pyridyl]carbamate
Formel: C₁₁H₁₄F₂N₂O₂
Molecular Weight: 244,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767513
CAS-Nummer: 1816284-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR