3-(Difluoromethyl)-2-iodoquinoline
Lieferant: Apollo Scientific
PC520030-1G
APOSPC520030-1GEA
3190
EUR
APOSPC520030-1G
3-(Difluoromethyl)-2-iodoquinoline
3-(Difluoromethyl)-2-iodoquinoline
Formel:
C₁₀H₆F₂IN Molecular Weight: 305.063 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767514 CAS-Nummer: 2091217-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...