6-Chloro-5-(difluoromethyl)-1-isopropyl-pyrazolo[3,4-b]pyridine

Lieferant: Apollo Scientific

PC520031-1G
APOSPC520031-1GEA 3650 EUR
APOSPC520031-1G
6-Chloro-5-(difluoromethyl)-1-isopropyl-pyrazolo[3,4-b]pyridine
6-Chloro-5-(difluoromethyl)-1-isopropyl-pyrazolo[3,4-b]pyridine
Formel: C₁₀H₁₀ClF₂N₃
Molecular Weight: 245,656 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767515
CAS-Nummer: 2090187-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR