2-Chloro-3-(difluoromethyl)-7-methoxyquinoline

Lieferant: Apollo Scientific

PC520032-1G
APOSPC520032-1GEA 2740 EUR
APOSPC520032-1G
2-Chloro-3-(difluoromethyl)-7-methoxyquinoline
2-Chloro-3-(difluoromethyl)-7-methoxyquinoline
Formel: C₁₁H₈ClF₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD29767516
CAS-Nummer: 2091217-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR